Order Entry
Belgium
ContactUsLinkComponent
Lithium standard solution, 1000 mg/L Li in dil. nitric acid (from Li₂CO₃) standard for AAS and ICP
Lithium standard solution, 1000 mg/L Li in dil. nitric acid (from Li₂CO₃) standard for AAS and ICP
Catalog # PERCN9300129
Supplier:  PerkinElmer
CAS Number:  
Lithium standard solution, 1000 mg/L Li in dil. nitric acid (from Li₂CO₃) standard for AAS and ICP
Catalog # PERCN9300129
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Lithium
  • Symbol:
    Li

Specifications

About this item