Order Entry
Belgium
ContactUsLinkComponent
Indium standard solution, 1,000 mg/L In in dilute nitric acid (from In) standard for AAS and ICP
Indium standard solution, 1,000 mg/L In in dilute nitric acid (from In) standard for AAS and ICP
Catalog # PERCN9300124
Supplier:  PerkinElmer
CAS Number:  
Indium standard solution, 1,000 mg/L In in dilute nitric acid (from In) standard for AAS and ICP
Catalog # PERCN9300124
Supplier:  PerkinElmer
CAS Number:  

Specifications

  • Concentration:
    1000 mg/L
  • Matrix:
    2% HNO₃
  • Pk:
    1
  • Element:
    Indium
  • Symbol:
    In

Specifications

About this item