Order Entry
Belgium
ContactUsLinkComponent
Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid (from Au) standard for AAS and ICP
Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid (from Au) standard for AAS and ICP
  PERCN9300121
 :  
Gold standard solution, 1,000 mg/L Au in 10% hydrochloric acid (from Au) standard for AAS and ICP
  PERCN9300121
 :  

 

  • Concentration:
    1000 mg/L
  • Matrix:
    10% HCl
  • Pk:
    1
  • Element:
    Gold
  • Symbol:
    Au