β-Nicotinamide adenine dinucleotid, oxidised form (NAD, oxidised form)
: Serva
:53-84-9
:
:C₂₁H₂₇N₇O₁₄P₂
:MFCD00036253
:663.43 g/mol
:13,06370
:140 to 142 °C
:Refrigerator
:200-184-4
Frequently Bought Together
VWR®, Straight Sample Containers, with Screw Cap
: VWR Collection
Eppendorf® Protein LoBind® Microcentrifuge Tubes
: EPPENDORF
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Acetone ≥99%, TECHNICAL
: VWR Chemicals
Transfer pipettes
: RATIOLAB
Petroleum spirit, 40…60 °C, GPR RECTAPUR®
: VWR Chemicals
Safe-Lock Tubes, 2,0 ml - Microtubes
: EPPENDORF
Water, HiPerSolv CHROMANORM® for HPLC
: VWR Chemicals
Safe-Lock Tubes, 1,5 ml - Microtubes
: EPPENDORF
VWR®, Spreaders, L-shaped, Disposable
: VWR Collection
VWR®, Transfer Pipettes, High Performance
: VWR Collection
VWR® Serological Pipette, Standard Line
: VWR Collection
VWR®, Centrifuge Tubes, Conical Bottom
: VWR Collection