Orders LinkContactUsLinkComponent
Tissue pulveriser, Bessman type
Tissue pulveriser, Bessman type
Tissue pulveriser, Bessman type
Certificates
Tissue pulveriser, Bessman type
3 Options Available Below
Product Details & Documents
The stainless steel Bessman tissue pulveriser consists of a two-component mortar with handles and a pestle, specifically designed for pulverising 10 - 1000 mg of tissue.
After chilling the pulveriser in liquid nitrogen, the included 28 cm lead hammer is used on the mortar to completely homogenise the inserted sample. The sample can then easily be transferred without touching or thawing by removing the mortar and inverting the pestle assembly over a test tube.
Delivery information: The small and medium pulveriser includes 10 test tubes and the large pulveriser includes one culture jar.
Product Family Options
Product Information
  • Capacity
    Ø ext.×length
    Availability
    Price
  • SPEC189470
    Bessman tissue pulveriser, small
    Bessman tissue pulveriser, small
    10 – 50 mg
    6,6×26,8 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 878,00 €
    878,00 €
    1 * 1 ST
       878,00 €
  • SPEC189475
    Bessman tissue pulveriser, medium
    Bessman tissue pulveriser, medium
    25 – 250 mg
    12,7×47,3 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 100,00 €
    1 100,00 €
    1 * 1 ST
       1 100,00 €
  • SPEC189476
    Bessman tissue pulveriser, large
    Bessman tissue pulveriser, large
    100 – 1000 mg
    25,6×47,6 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 040,00 €
    1 040,00 €
    1 * 1 ST
       1 040,00 €