Orders LinkContactUsLinkComponent
Tabletop reaction units, labscale
Tabletop reaction units, labscale
Supplier:   Lenz Laborglas
Tabletop reaction units, labscale
Supplier:  Lenz Laborglas
Certificates
Tabletop reaction units, labscale
Tabletop reaction units, labscale
6 Options Available Below
Product Details & Documents
These table top reaction units are used for stirring under normal pressure or vacuum, with possibility of addition of liquids and boiling under reflux.
High quality system components include:
Reaction Vessels (100, 250, 500, 1000 or 2000 ml): made of DURAN® tubing, with thermostatic jacket and bottom discharge valve.
Flat flange lid (LF 60 or LF 100): DURAN®, with center neck and two angled side necks.
Cylindrical funnels (100 or 250 ml): DURAN® tubing, with graduation.
Reflux condensers: made of DURAN® tubing.
Stirrer guide and stirrer: with 10 mm stirrer shaft and lateral PTFE blade.
Flat flange lid (LF 60 or LF 100): DURAN®, with center neck and two angled side necks.
Cylindrical funnels (100 or 250 ml): DURAN® tubing, with graduation.
Reflux condensers: made of DURAN® tubing.
Stirrer guide and stirrer: with 10 mm stirrer shaft and lateral PTFE blade.
Product Family Options
Product Information
  • Availability
    Price
  • LENZ06260137-V2
    100 ml, LF 60
    100 ml, LF 60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 510,00 €
    2 510,00 €
    1 * 1 ST
       2 510,00 €
  • LENZ06260149-V2
    250 ml, LF 60
    250 ml, LF 60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 520,00 €
    2 520,00 €
    1 * 1 ST
       2 520,00 €
  • LENZ06260158-V2
    500 ml, LF 60
    500 ml, LF 60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 530,00 €
    2 530,00 €
    1 * 1 ST
       2 530,00 €
  • LENZ06260258-V2
    500 ml, LF 100
    500 ml, LF 100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 990,00 €
    2 990,00 €
    1 * 1 ST
       2 990,00 €
  • LENZ06260270-V2
    1000 ml, LF 100
    1000 ml, LF 100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 010,00 €
    3 010,00 €
    1 * 1 ST
       3 010,00 €
  • LENZ06260273-V2
    2000 ml, LF 100
    2000 ml, LF 100
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 030,00 €
    3 030,00 €
    1 * 1 ST
       3 030,00 €
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Support, complete with clamps and connectors, for LF 60
    Support, complete with clamps and connectors, for LF 60
    LENZ06800000
    Log in for availability
    1 * 1 ST - 181,00 €