Digital Solutions
Applications
Support
About
Belgium
ContactUsLinkComponent
Pipette Tips
Pipette tips for Assipette pipettes
Pipette tips for Assipette pipettes
HECH40110010
:
GLASWARENFABRIK KARL HECHT
Pipette tips for Assipette pipettes
HECH40110010
:
GLASWARENFABRIK KARL HECHT
Packed:
Bag of 1000
Volume:
5 - 100 µl
Tip colour:
Yellow
For use with:
Assipettes, Brand, Eppendorf, La Fontaine, Gilson, Lab Systems piston pipettes
Pk:
1.000