Orders LinkContactUsLinkComponent
Blanking nuts
Blanking nuts
 :   COWIE
Blanking nuts
 :  COWIE
Blanking nuts
Blanking nuts
PTFE, white.
  • Chemically inert
  • Reduce contamination and simplify cleaning
  • Non-scratch
NPT blanking nuts/plugs are used to seal unused ports. To assemble simply screw the NPT blanking nut into a corresponding NPT port of the reactor lid.
Product Family Options
  • Description
  • fit port size ¼" NPT
    1 * 1 ST
    1 * 1 ST - 6,80 €
    6,80 €
    1 * 1 ST
       6,80 €
  • fit port size ½" NPT
    1 * 1 ST
    1 * 1 ST - 7,00 €
    7,00 €
    1 * 1 ST
       7,00 €
  • fit port size ¾" NPT
    1 * 1 ST
    1 * 1 ST - 10,80 €
    10,80 €
    1 * 1 ST
       10,80 €
  • fit port size 1" NPT
    1 * 1 ST
    1 * 1 ST - 14,90 €
    14,90 €
    1 * 1 ST
       14,90 €
  • fit port size 1½" NPT
    1 * 1 ST
    1 * 1 ST - 28,30 €
    28,30 €
    1 * 1 ST
       28,30 €