Blanking lids with handle, pure PTFE
Supplier: COWIE
Jump to:
About this item
PTFE, white.
- Chemically inert
- Designed to reduce contamination and simplify cleaning
- Non-Scratch
- Reduced breakages
- Improved safety
Blanking lids have no ports and are designed to provide a seal for reaction vessels. Lids fit 60 mm, 100 mm, 150 mm, 200 mm Schott® type flanges and have a central handle to facilitate easy handling.
4 Options Available Below
Product Details & Documents
PTFE, white.
- Chemically inert
- Designed to reduce contamination and simplify cleaning
- Non-Scratch
- Reduced breakages
- Improved safety
Blanking lids have no ports and are designed to provide a seal for reaction vessels. Lids fit 60 mm, 100 mm, 150 mm, 200 mm Schott® type flanges and have a central handle to facilitate easy handling.
Product Family Options
Product Information
- WithExt.ØTypeTotal heightAvailabilityPrice