Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cohesive sampler

Cohesive sampler

Cohesive sampler, Stainless steel 316
Catalog # SAMP2000S-01
Supplier:   Sampling Systems
Cohesive sampler
Catalog # SAMP2000S-01
Supplier:  Sampling Systems
Cohesive sampler
Cohesive sampler

Cohesive sampler, Stainless steel 316

Price
1 370,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

This cohesive sampler is designed for powders.

  • For taking single point samples
  • Handle and four sample cells
  • 0,25, 0,50, 0,75, 1,0 ml unit doses

Used throughout the pharmaceutical industry from sticky (or cohesive) powders.

Ordering information: A Nylon bristle can be ordered separately.

Delivery information: In a case.

Specifications
  • Description:
    Cohesive sampler
  • Material:
    Stainless steel 316
Accessories

Product Information

  • Catalog #
    Bristle material
    Brush head Ø
    Handle material
    Availability
    Price
  • 300-0513
    Nylon
    7 - 14 mm
    Wire
    Log in for availability
    1 * 1 ST - 49,50 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.