ContactUsLinkComponent
Hydrophilic interaction chromatography columns, Asahipak NH2P preparative type
Hydrophilic interaction chromatography columns, Asahipak NH2P preparative type
Supplier:  Resonac
Certificates
Hydrophilic interaction chromatography columns, Asahipak NH2P preparative type
Supplier:  Resonac
Specifications
  • Catalog No:
  • 554-5307
  • 554-5308
  • 554-5306
  • Description:
  • Asahipak NH2P-90 20F
  • Asahipak NH2P-130G 7B
  • Asahipak NH2P-50 10E
  • Max. pressure:
  • 10 MPa (100 bar)
  • 15 MPa (150 bar)
  • pH range:
  • 2 - 13
  • 2 - 13
  • 2 - 13
  • Temperature limit:
  • 4 - 50 °C
  • 4 - 50 °C
  • 4 - 50 °C
  • USP code:
  • L82
  • L82
  • L82