Orders LinkContactUsLinkComponent
Sleeve Sampler
Sleeve Sampler
Supplier:   Sampling Systems
Sleeve Sampler
Supplier:  Sampling Systems
Certificates
Sleeve Sampler
6 Options Available Below
Product Details & Documents
The sleeve sampler is specially designed for large volumes at great depths.
  • Adjustable length and bright finish design
  • Extremely strong and robust extension rods
Suitable for free-flowing powders and granules.
Product Family Options
Product Information
  • Material
    Length
    Availability
    Price
  • SAMP1035T-150
    Sample chamber, 150 ml
    Sample chamber, 150 ml
    Stainless steel 316L
    1500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 410,00 €
    410,00 €
    1 * 1 ST
       410,00 €
  • SAMP1035P-150
    Sample chamber, 150 ml
    Sample chamber, 150 ml
    Stainless steel 316L
    1500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 751,00 €
    751,00 €
    1 * 1 ST
       751,00 €
  • SAMP1035T-200
    Sample chamber, 200 ml
    Sample chamber, 200 ml
    Stainless steel 316L
    200 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 416,00 €
    416,00 €
    1 * 1 ST
       416,00 €
  • SAMP1035A-1000
    Heavy duty handle
    Heavy duty handle
    Stainless steel 316L
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 292,00 €
    292,00 €
    1 * 1 ST
       292,00 €
  • SAMP1035A-1500
    Heavy duty handle
    Heavy duty handle
    Stainless steel 316L
    1500 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 324,00 €
    324,00 €
    1 * 1 ST
       324,00 €
  • SAMP1035E-1000
    Extension rod
    Extension rod
    Stainless steel 316L
    1000 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 282,00 €
    282,00 €
    1 * 1 ST
       282,00 €