ContactUsLinkComponent
Syringe filters, Puradisc™ Aqua 30, Whatman™
undefined
Syringe filters, Puradisc™ Aqua 30, Whatman™
  • Catalog No:
  • 514-0102
  • 514-0288
  • Dimensions:
  • 26×34 mm
  • 26×34 mm
  • Max. operating pressure:
  • 6,9 bar
  • 6,9 bar
  • Weight:
  • 4,7 g
  • 4,7 g
Frequently Bought Together