Redox/ORP electrodes
Supplier: CONSORT
Jump to:
About this item
2 Options Available Below
Product Details & Documents
Product Family Options
Product Information
- TypeConnectionMaterialTemp.Ø×LForAvailabilityPrice
- CONSSO50XPlatinum ORP (redox) electrode, 0...±2000 mVPlatinum ORP (redox) electrode, 0...±2000 mVSingle junction, sealedS7Epoxy0...+80 °C12×110 mmGeneral purposesLog in for availability1 * 1 ST190,00 €
- CONSSO60XPlatinum ORP (redox) electrode, 0...±2000 mVPlatinum ORP (redox) electrode, 0...±2000 mVSingle junction, refillableS7Glass0...+100 °C12×110 mmGeneral purposesLog in for availability1 * 1 ST184,00 €