Orders LinkContactUsLinkComponent
Filter sets
Filter sets
Supplier:   ZEISS
Filter sets
Supplier:  ZEISS
Certificates
13 Options Available Below
Product Details & Documents
Product Family Options
Product Information
  • Availability
    Price
  • ZEIS4880029901000
    Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
    Filter set 02, excitation G 365, beam splitter FT 395, emission LP 420
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 313,00 €
    1 313,00 €
    1 * 1 ST
       1 313,00 €
  • ZEIS4880099901000
    Filter set 09, excitation BP 450 - 490, beam splitter FT 510, emission LP 515
    Filter set 09, excitation BP 450 - 490, beam splitter FT 510, emission LP 515
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 848,86 €
    1 848,86 €
    1 * 1 ST
       1 848,86 €
  • 630-1422
    Filter set 15, excitation BP 546/12, beam splitter FT 580, emission LP 590
    Filter set 15, excitation BP 546/12, beam splitter FT 580, emission LP 590
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 662,29 €
    1 662,29 €
    1 * 1 ST
       1 662,29 €
  • ZEIS4880209901000
    Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
    Filter set 20, excitation BP 546/12, beam splitter FT 560, emission BP 575-640
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 747,86 €
    1 747,86 €
    1 * 1 ST
       1 747,86 €
  • ZEIS4880250000000
    Filter set 25, excitation TBP 400 + 495 + 570, beam splitter TFT 410 + 505 + 585, emission TBP 460 + 530 + 625
    Filter set 25, excitation TBP 400 + 495 + 570, beam splitter TFT 410 + 505 + 585, emission TBP 460 + 530 + 625
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 3 654,24 €
    3 654,24 €
    1 * 1 SET
       3 654,24 €
  • ZEIS0000001031350
    Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60
    Filter set 31, excitation BP 565/30, beam splitter FT 585, emission BP 620/60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 754,87 €
    1 754,87 €
    1 * 1 ST
       1 754,87 €
  • ZEIS0000001031346
    Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
    Filter set 38, excitation BP 470/40, beam splitter FT 495, emission BP 525/50
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 754,87 €
    1 754,87 €
    1 * 1 ST
       1 754,87 €
  • ZEIS4890389901000
    Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
    Filter set 38 (high efficiency), excitation BP 470/40 (HE), beam splitter FT 495 (HE), emission BP 525/50 (HE)
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 456,26 €
    2 456,26 €
    1 * 1 ST
       2 456,26 €
  • ZEIS0000001114101
    Filter set 43, excitation BP 545/25, beam splitter FT 570, emission BP 605/70
    Filter set 43, excitation BP 545/25, beam splitter FT 570, emission BP 605/70
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 1 745,06 €
    1 745,06 €
    1 * 1 SET
       1 745,06 €
  • ZEIS0000001114459
    Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
    Filter set 44, excitation BP 475/40, beam splitter FT 500, emission BP 530/50
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 1 745,06 €
    1 745,06 €
    1 * 1 ST
       1 745,06 €
  • ZEIS4880459100400
    Filter set 45, excitation BP 560/40, beam splitter FT 585, emission BP 630/75
    Filter set 45, excitation BP 560/40, beam splitter FT 585, emission BP 630/75
    Log in for availability
    Log in for availability
    1 * 1 Pce
    1 * 1 Pce - 1 745,06 €
    1 745,06 €
    1 * 1 Pce
       1 745,06 €
  • ZEIS4890630000000
    Filter set 63 (high efficiency), excitation BP 572/25 (HE), beam splitter FT 590 (HE), emission BP 629/62 (HE)
    Filter set 63 (high efficiency), excitation BP 572/25 (HE), beam splitter FT 590 (HE), emission BP 629/62 (HE)
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET - 2 421,19 €
    2 421,19 €
    1 * 1 SET
       2 421,19 €
  • ZEIS4890969100400
    Filter set 96 HE with reflector module FL EC P and C (E), consisting of the filters EX BP 390/40, BS FT 420, EM BP 450/40 installed in reflector module with reflection prevention through inclined mount for emission filter
    Filter set 96 HE with reflector module FL EC P and C (E), consisting of the filters EX BP 390/40, BS FT 420, EM BP 450/40 installed in reflector module with reflection prevention through inclined mount for emission filter
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 687,72 €
    2 687,72 €
    1 * 1 ST
       2 687,72 €