ContactUsLinkComponent
Cation exchange chromatography columns, IEC series
Cation exchange chromatography columns, IEC series
Supplier:  Resonac
Certificates
Cation exchange chromatography columns, IEC series
Supplier:  Resonac
Certificates
Specifications
  • Catalog No:
  • 554-2856
  • 554-2851
  • Max. pressure:
  • 2,0 MPa (20 bar)
  • 2,0 MPa (20 bar)
  • pH range:
  • 2 - 12
  • 2 - 12
  • Temperature limit:
  • 10 - 50 °C
  • 10 - 50 °C