Orders LinkContactUsLinkComponent
Ion selective electrodes
Ion selective electrodes
Supplier:   CONSORT
Ion selective electrodes
Supplier:  CONSORT
Certificates
Ion selective electrodes
Ion selective electrodes
8 Options Available Below
Product Details & Documents
These combination electrodes have an epoxy body.
Solid state electrodes utilise relatively insoluble inorganic salts in a membrane. Solid state electrodes exist in homogeneous or heterogeneous forms. In both types, potentials are developed at the membrane surface due to the ion-exchange process.
Glass membrane electrodes are formed by the doping of the silicon dioxide glass matrix with various chemicals. The most common of the glass membrane electrodes is the pH electrode. Glass membrane electrodes are also available for the measurement of sodium ions.
Polymer membrane electrodes consist of various ionexchange materials in an inert matrix such as PVC, polythene or silicone rubber. The potential developed at the membrane surface is related to the concentration of the species of interest.
Documents
Product Family Options
Product Information
  • Concentration
    pH
    Temp.
    Type
    Material
    Electrolyte
    Connection
    Ø×L
    Availability
    Price
  • CONSISE23B
    Combination electrode, Calcium (Ca²⁺)
    Combination electrode, Calcium (Ca²⁺)
    5,10⁻⁶ - 10° mol/l; 0,2 - 40000 mg/l
    3 - 10
    0...+50 °C
    Polymer sensor
    Epoxy
    KCl
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE24B
    Combination electrode, Chloride (Cl⁻)
    Combination electrode, Chloride (Cl⁻)
    5,10⁻⁵ - 10° mol/l; 1,8 - 35500 mg/l
    1 - 12
    0...+50 °C
    Solid state sensor
    Epoxy
    KNO₃
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE25B
    Combination electrode, Copper (Cu²⁺)
    Combination electrode, Copper (Cu²⁺)
    10⁻⁸ - 10⁻¹ mol/l; 0,00064 - 6350 mg/l
    2 - 12
    0...+50 °C
    Solid state sensor
    Epoxy
    KNO₃
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE26B
    Combination electrode, Copper (CN⁻)
    Combination electrode, Copper (CN⁻)
    5,10⁻⁶ - 10⁻² mol/l; 0,13 - 260 mg/l
    11 - 13
    0...+50 °C
    Solid state sensor
    Epoxy
    KNO₃
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE27B
    Combination electrode, Copper (F⁻)
    Combination electrode, Copper (F⁻)
    10⁻⁶ - sat. mol/l; 0,02 mg/l - sat.
    5 - 8
    0...+50 °C
    Solid state sensor
    Epoxy
    KCl
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE33B
    Combination electrode, Potassium (K⁺)
    Combination electrode, Potassium (K⁺)
    10⁻⁶ - 10° mol/l; 0,04 - 39000 mg/l
    2 - 12
    0...+50 °C
    Polymer sensor
    Epoxy
    NaCl
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE34B
    Combination electrode, Silver/Sulphide (Ag⁺/S2⁻)
    Combination electrode, Silver/Sulphide (Ag⁺/S2⁻)
    10⁻⁷ - 10° mol/l; 0,01 - 107900 mg/l; 0,003 - 32000 mg/l
    2 - 12
    0...+50 °C
    Solid state sensor
    Epoxy
    KNO₃
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
  • CONSISE35B
    Combination electrode, Sodium (Na⁺)
    Combination electrode, Sodium (Na⁺)
    10⁻⁶ - sat. mol/l; 0,02 mg/l - sat.
    5 - 12
    0...+50 °C
    Glass sensor
    Epoxy
    NH₄Cl
    BNC, 1 m cable
    12×110 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 957,00 €
    957,00 €
    1 * 1 ST
       957,00 €
Recommandations
Les recommandations seront personnalisées en fonction de vos préférences d’achat uniquement si vous avez donné votre consentement en activant l’option " Améliorer mon expérience d’achat " sur la page "Mon compte".
Dans le cas contraire, vous recevrez des recommandations génériques.