Conductivity/TDS/°C meter, bench, K912
Supplier: CONSORT
Certificates
About this item
Robust, dust- and splash-proof bench meter with one input for a 4-pole conductivity electrode with built-in Pt1000 sensor.
- High resolution graphic screen with white backlight indicates all measurements and operating parameters such as date and time
- Multi-point (1 - 3) conducticity calibration for more linearity
- Works with a 4-pole electrode eliminating polarisation effects and electrode fouling
- Storage memory for up to 3600 values + date and time
- Programmable recorder output, two-way communication with any computer using RS232
Delivery information: The kits are supplied with conductivity ATC electrode SK40T (epoxy electrode), 3×50 ml conductivity standards (0,01, 0,1 and 1 M KCl), batteries, mains adapter and carrying case.
Frequently Bought Together