Order Entry
Belgium
Orders LinkContactUsLinkComponent
Colour Reference solution, CertiPUR®, Supelco®

Colour Reference solution, CertiPUR®, Supelco®

Hazen 500 standard, Platin/cobalt reference solution, CertiPUR®, Glass bottle, 250 mL
Catalog # 1.00246.0250
Supplier:   Merck
Colour Reference solution, CertiPUR®, Supelco®
Catalog # 1.00246.0250
Supplier:  Merck
CAS Number:  
Colour Reference solution, CertiPUR®, Supelco®
Colour Reference solution, CertiPUR®, Supelco®

Hazen 500 standard, Platin/cobalt reference solution, CertiPUR®, Glass bottle, 250 mL

CAS Number:  
Price
258,00 €
Log in for availability
1 * 250 mL
Product Details & Documents

The colour reference solution can be used in visual comparison or as part of a calibration of colorimeters and spectrophotometer for colour comparison. The solution can be used to prepare reference solution with different Hazen values.

  • Traceable to NIST SRM930e
  • Shelf life of unopened bottle is minimum of 18 months

For use under DIN EN ISO 7887, ASTM D 1209-05, DIN EN ISO 6271-1, APHA 2120 B & C

Documents

Certificates
Search by Lot Number
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    250 mL
  • Description:
    Hazen 500 standard, Platin/cobalt reference solution, CertiPUR®

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.