Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Furniture
Shelves
Shelf for compact 18 L heratherm incubator
Shelf for compact 18 L heratherm incubator
Shelf for compact 18 L heratherm incubator
Catalog # 466-0311
Supplier:
Thermo Fisher Scientific
Expand image
Shelf for compact 18 L heratherm incubator
Catalog # 466-0311
Supplier:
Thermo Fisher Scientific
Specifications
Description:
Perforated stainless steel shelf
Pk:
1
For:
IMC18 Compact 18 litre incubator
Specifications
About this item
Product Details
Product Details