Conductivity electrodes
Supplier: CONSORT
Jump to:
About this item
A range of conductivity electrodes for different applications.
- Available with or without integrated temperature sensor
- Shaft is made from glass or epoxy
7 Options Available Below
Product Details & Documents
A range of conductivity electrodes for different applications.
- Available with or without integrated temperature sensor
- Shaft is made from glass or epoxy
Product Family Options
Product Information
- TypeCell constantTemp.MaterialØ×LConnectionForAvailabilityPrice
- CONSSK10BConductivity sensor, dual graphite platesConductivity sensor, dual graphite platesStandard1 cm⁻¹0...80 °CEpoxy12×110 mmBNC + 2 banana, 1 m cableGeneral purposes, education, agricultureLog in for availability1 * 1 ST101,00 €
- CONSSK10TConductivity sensor, dual graphite plates, with built-in ATC (Pt1000)Conductivity sensor, dual graphite plates, with built-in ATC (Pt1000)Standard, with temp. sensor1 cm⁻¹0...80 °CEpoxy12×110 mmBNC + 2 banana, 1 m cableGeneral purposes, education, agricultureLog in for availability1 * 1 ST110,00 €
- CONSSK20BConductivity sensor, dual plantinum ringsConductivity sensor, dual plantinum ringsStandard1 cm⁻¹0...110 °CGlass12×110 mmBNC + 2 banana, 1 m cableGeneral purposes, research, laboratoryLog in for availability1 * 1 ST165,00 €
- CONSSK20TConductivity sensor, dual plantinum rings, with built-in ATC (Pt1000)Conductivity sensor, dual plantinum rings, with built-in ATC (Pt1000)Standard, with temp. sensor1 cm⁻¹0...110 °CGlass12×110 mmBNC + 2 banana, 1 m cableGeneral purposes, research, laboratoryLog in for availability1 * 1 ST225,00 €
- CONSSK21Tsensor for low conductivities, dual plantinum plates, with built-in ATC (Pt1000)sensor for low conductivities, dual plantinum plates, with built-in ATC (Pt1000)Low conductivity sensor, with temp. sensor0,1 cm⁻¹0...110 °CGlass12×110 mmBNC + 2 banana, 1 m cableLow conductivities, distilled waterLog in for availability1 * 1 ST257,00 €
- CONSSK23Tsensor for high conductivities, dual plantinum rings, with built-in ATC (Pt1000)sensor for high conductivities, dual plantinum rings, with built-in ATC (Pt1000)High conductivity sensor, with temp. sensor10 cm⁻¹0...110 °CGlass12×130 mmBNC + 2 banana, 1 m cableHigh conductivites, sea waterLog in for availability1 * 1 ST257,00 €
- CONSSK24TMicro sensor, dual plantinum plates, with built-in ATC (Pt 1000)Micro sensor, dual plantinum plates, with built-in ATC (Pt 1000)Micro, with temp. sensor1 cm⁻¹0...100 °CGlass6×110 mmBNC + 2 banana, 1 m cableMicro volumes (min. 30 mm depth), test tubesLog in for availability1 * 1 ST482,00 €