Orders LinkContactUsLinkComponent
Twaddle hydrometers
Supplier:   Alla France
Twaddle hydrometers
Supplier:  Alla France
Certificates
Twaddle hydrometers
Twaddle hydrometers
4 Options Available Below
Product Details & Documents
Hydrometers for sugar with scales.
Calibration temperature of 15.6 °C.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • 614-1622
    Twaddle hydrometer
    Twaddle hydrometer
    24 – 48 °TW
    0,5 °TW
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,40 €
    34,40 €
    1 * 1 ST
       34,40 €
  • 614-1623
    Twaddle hydrometer
    Twaddle hydrometer
    48 – 74 °TW
    0,5 °TW
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,90 €
    49,90 €
    1 * 1 ST
       49,90 €
  • 614-1624
    Twaddle hydrometer
    Twaddle hydrometer
    74 – 102 °TW
    0,5 °TW
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,40 €
    34,40 €
    1 * 1 ST
       34,40 €
  • 614-1625
    Twaddle hydrometer
    Twaddle hydrometer
    102 – 138 °TW
    0,5 °TW
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 57,40 €
    57,40 €
    1 * 1 ST
       57,40 €