Orders LinkContactUsLinkComponent
Hydrometers, Brix
Hydrometers, Brix
Supplier:   Alla France
Hydrometers, Brix
Supplier:  Alla France
Certificates
Hydrometers, Brix
Hydrometers, Brix
3 Options Available Below
Product Details & Documents
These Brix sugar scale hydrometers are specially designed for use in breweries.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • 0 - 30 °Bx
    0,5 °Bx
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,90 €
    36,90 €
    1 * 1 ST
       36,90 €
  • 30 - 60 °Bx
    0,1 °Bx
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,50 €
    38,50 €
    1 * 1 ST
       38,50 €
  • 60 - 90 °Bx
    0,1 °Bx
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 55,80 €
    55,80 €
    1 * 1 ST
       55,80 €