Orders LinkContactUsLinkComponent
Hydrometers
Hydrometers
Supplier:   Alla France
Hydrometers
Supplier:  Alla France
Certificates
Hydrometers
4 Options Available Below
Product Details & Documents
Glass hydrometer calibrated at 60 °F (15,56 °C).
  • Graduated in specific gravity (SG)
  • Reading below meniscus
Packaging: In a plastic case.
Services: Other temperatures of calibration upon request.
Product Family Options
Product Information
  • Range
    Division
    Length
    Material
    Availability
    Price
  • Hydrometer
    0,650 – 1,000 SG
    0,005 SG
    310 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,70 €
    21,70 €
    1 * 1 ST
       21,70 €
  • 614-1642
    Hydrometer
    Hydrometer
    0,800 – 1,000 SG
    0,002 SG
    300 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,50 €
    31,50 €
    1 * 1 ST
       31,50 €
  • Hydrometer
    0,650 – 1,000 SG
    0,01 SG
    290 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 49,70 €
    49,70 €
    1 * 1 ST
       49,70 €
  • Hydrometer
    1,500 – 2,000 SG
    0,005 SG
    235 mm
    Glass
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 47,60 €
    47,60 €
    1 * 1 ST
       47,60 €