Orders LinkContactUsLinkComponent
Soil hydrometers
Soil hydrometers
Supplier:   Alla France
Soil hydrometers
Supplier:  Alla France
Certificates
Soil hydrometers
Soil hydrometers
3 Options Available Below
Product Details & Documents
Soil hydrometers for specific gravity (SG) measurements.
  • Medium surface tension (55 mN/m)
  • Reading below meniscus
Certifications: ASTM E100 D422 - A2LA traceable.
Ordering information: EU traceable calibration certificate or EO official calibration certificate available on request.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • 614-1534
    Soil hydrometer
    Soil hydrometer
    0,995 − 1,038 S.G.
    0,001 S.G.
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,80 €
    83,80 €
    1 * 1 ST
       83,80 €
  • ALLA3470FC06068QP
    Soil hydrometer
    Soil hydrometer
    −5 − +60 g/l
    1000 g/l
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 83,80 €
    83,80 €
    1 * 1 ST
       83,80 €
  • ALLA3470FC06068EU
    Soil hydrometer with three-point calibration certificate
    Soil hydrometer with three-point calibration certificate
    −5 − +60 g/l
    1000 g/l
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 498,00 €
    498,00 €
    1 * 1 ST
       498,00 €