Order Entry
Belgium
Orders LinkContactUsLinkComponent
Weigh boxes, XPert™

Weigh boxes, XPert™

HEPA filtered weigh box, XPert™, with 2 glove ports, incl. a pair of neoprene gloves
Catalog # 135-1097
Supplier:   LABCONCO
Weigh boxes, XPert™
Catalog # 135-1097
Supplier:  LABCONCO
Weigh boxes, XPert™
Weigh boxes, XPert™

HEPA filtered weigh box, XPert™, with 2 glove ports, incl. a pair of neoprene gloves

Price
18 990,00 €
Log in for availability
1 * 1 ST
Product Details & Documents

XPert weigh boxes have inlet and outlet HEPA filters and provide a leak-tight physical barrier to protect the operator from hazardous airborne particulates and powders.

  • One-piece molded, medium-density PE shell; dry powder epoxy-coated steel superstructure
  • 1/4" thick laminated safety glass viewing window, removable for loading equipment, with powder-coated steel frame and neoprene gasket
  • Space-saving inner and outer transfer chamber doors that pivot upward, are counterbalanced and equipped with quick-latches
  • Black, chemical-resistant work surface sealed to the glove box floor
  • Anti-static ionizer fan to circulate ionized air inside the box to reduce static charge during weighing
  • Built-in blower with speed control capable of airflow from 13 CFM to 100 CFM
  • Inlet and outlet HEPA filters, 99,99% efficient on 0,3 micron particulates

These boxes include an anti-static ionizer fan to reduce static charge and a balance vibration isolator, both useful during weighing operations. Airborne particulate cleanliness (measured under positive and negative pressures per ISO 14644-1 test method) exceeds ISO Class 5 conditions (equivalent to Class 100) and achieved ISO Class 3 conditions (equivalent to Class 1) at both minimum and maximum operational airflow volumes.

Certifications: Meet Class 1 atmosphere containment conditions for oxygen leak rates per ISO 10648-2 test method

Delivery information: Supplied with 1 pair of neoprene gloves (size 9 3/4").

Documents

Specifications
  • Description:
    HEPA filtered weigh box, XPert™, with 2 glove ports, incl. a pair of neoprene gloves
  • Cat. No.:
    135-1097
  • W×D×H int.:
    851×698×635 mm
  • W×D×H ext.:
    1340×800×1020 mm
  • Observation window (W×H):
    670×317,5 mm
  • Volume:
    368 L
  • Model:
    XPert™
  • Transfer chamber W×D×H:
    Ø 280 × 305 mm
  • Weight:
    159 kg
Accessories

Product Information

  • Catalog #
    Availability
    Price
  • Thimble exhaust connector, Ø 12,7 cm, for connecting the glove box to the FilterMate™ portable exhauster
    Thimble exhaust connector, Ø 12,7 cm, for connecting the glove box to the FilterMate™ portable exhauster
    135-1090
    Log in for availability
    1 * 1 ST - 513,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Mechanical or electrical pass-through kit, provides three pass-through ports with sealing ID ranges 0,230/0,395, 0,230/0,530 and 0,450/0,705, useful for sealing cords and plumbing (Requires installation and drill holes; installation kit 5240800 is recommended)
    Mechanical or electrical pass-through kit, provides three pass-through ports with sealing ID ranges 0,230/0,395, 0,230/0,530 and 0,450/0,705, useful for sealing cords and plumbing (Requires installation and drill holes; installation kit 5240800 is recommended)
    LBCP5240700
    Log in for availability
    1 * 1 ST - 62,50 €

Product Information

  • Catalog #
    Availability
    Price
  • Bag set for bag-in/bag-out HEPA filter replacement, incl. two bags and two straps necessary for changing and disposing of one inlet or outlet filter
    Bag set for bag-in/bag-out HEPA filter replacement, incl. two bags and two straps necessary for changing and disposing of one inlet or outlet filter
    LBCP5241700
    Log in for availability
    1 * 1 ST - 478,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Blower foot switch, incl. foot pedal and power cord; provides a hands-free means to temporarily shut off power when obtaining sensitive balance readings or manipulating fine powders
    Blower foot switch, incl. foot pedal and power cord; provides a hands-free means to temporarily shut off power when obtaining sensitive balance readings or manipulating fine powders
    135-1089
    Log in for availability
    1 * 1 ST - 611,00 €

Product Information

  • Catalog #
    Availability
    Price
  • DOP test kit for HEPA filters, incl. the connections, hoses and test procedure necessary to conduct a HEPA filter test on both the inlet and outlet filter
    DOP test kit for HEPA filters, incl. the connections, hoses and test procedure necessary to conduct a HEPA filter test on both the inlet and outlet filter
    LBCP5242400
    Log in for availability
    1 * 1 ST - 301,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Installation kit for mechanical or electrical straight pass-through kit, incl. drill bits for installing the mechanical or electrical pass-through
    Installation kit for mechanical or electrical straight pass-through kit, incl. drill bits for installing the mechanical or electrical pass-through
    LBCP5240800
    Log in for availability
    1 * 1 KIT - 147,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Transfer chamber pressure gauge kit, incl. gauge, hose, mounting brackets, fittings and hardware; monitors pressure/vacuum from 0 to -29 inches of mercury (Requires Ø 0,562" drill hole in back of transfer chamber)
    Transfer chamber pressure gauge kit, incl. gauge, hose, mounting brackets, fittings and hardware; monitors pressure/vacuum from 0 to -29 inches of mercury (Requires Ø 0,562" drill hole in back of transfer chamber)
    LBCP5238100
    Log in for availability
    1 * 1 KIT - 552,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Recirculation kit, incl. hose, clamps and epoxy-coated steel adapter, allows the built-in blower exhaust to be connected to the inlet HEPA filter, providing total recirculation of air volume useful for dry atmosphere operations
    Recirculation kit, incl. hose, clamps and epoxy-coated steel adapter, allows the built-in blower exhaust to be connected to the inlet HEPA filter, providing total recirculation of air volume useful for dry atmosphere operations
    135-1086
    Log in for availability
    1 * 1 KIT - 198,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Positive pressure conversion kit, incl. hose, two clamps and harness assembly, creates particulate cleanliness inside the box measured to ISO 14644-1 test method, achieving ISO Class 3 conditions
    Positive pressure conversion kit, incl. hose, two clamps and harness assembly, creates particulate cleanliness inside the box measured to ISO 14644-1 test method, achieving ISO Class 3 conditions
    135-1087
    Log in for availability
    1 * 1 KIT - 311,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Thimble exhaust connector, Ø 15,2 cm, for connecting the glove box to ductwork attached to building exhaust, remote blower or FilterMate™ Portable Exhauster
    Thimble exhaust connector, Ø 15,2 cm, for connecting the glove box to ductwork attached to building exhaust, remote blower or FilterMate™ Portable Exhauster
    135-1084
    Log in for availability
    1 * 1 ST - 513,00 €

Product Information

  • Catalog #
    Availability
    Price
  • Hard duct exhaust connector, Ø 15,2 cm, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
    Hard duct exhaust connector, Ø 15,2 cm, for connecting the glove box to ductwork attached to a remote blower (Requires ductwork and remote blower)
    LBCP5230400
    Log in for availability
    1 * 1 ST - 513,00 €

Product Information

  • Catalog #
    Availability
    Price
  • ULPA filter, 99,999% efficiency (Ø 0,12 micron particles), may be used in place of standard inlet or outlet HEPA filter, weight 3 kg
    ULPA filter, 99,999% efficiency (Ø 0,12 micron particles), may be used in place of standard inlet or outlet HEPA filter, weight 3 kg
    LBCP5225106
    Log in for availability
    1 * 1 ST - 882,00 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.