Adapters, PTFE, socket on cone
Supplier: COWIE
Jump to:
About this item
Made of pure PTFE, white.
- Very good chemical resistance
- Very good temperature resistance
- Fully adjustable with replacement Viton® seal
- Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
12 Options Available Below
Product Details & Documents
Made of pure PTFE, white.
- Very good chemical resistance
- Very good temperature resistance
- Fully adjustable with replacement Viton® seal
- Contact materials are glass and PTFE only
This holder enables standard probes and thermometers to be assembled in standard taper laboratory glassware.
Product Family Options
Full length A type
Product Information
- TypeCone sizeSocket sizeAvailabilityPrice
This product has been blocked by your organization. Please contact your purchasing department for more information
Medium length B type
Product Information