Orders LinkContactUsLinkComponent
Extraction thimbles
Extraction thimbles
Extraction thimbles
Extraction thimbles
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
Glass tubes with sintered filter disc (porosity: 0), for extractors acc. to Thielepape.
Product Family Options
  • Capacity
  • 30 ml
    1 * 1 ST
    1 * 1 ST - 47,70 €
    47,70 €
    1 * 1 ST
       47,70 €
  • 150 ml
    1 * 1 ST
    1 * 1 ST - 49,40 €
    49,40 €
    1 * 1 ST
       49,40 €
  • 250 ml
    1 * 1 ST
    1 * 1 ST - 62,10 €
    62,10 €
    1 * 1 ST
       62,10 €