Orders LinkContactUsLinkComponent
Adapters, expansion
Adapters, expansion
Supplier:   witeg Labortechnik
Adapters, expansion
Supplier:  witeg Labortechnik
Certificates
Adapters, expansion
Adapters, expansion
3 Options Available Below
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapters.
Certifications: According to DIN 12257.
Product Family Options
Product Information
  • Type
    Cone size
    Socket size
    Availability
    Price
  • Expansion
    10/19 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,70 €
    18,70 €
    1 * 1 ST
       18,70 €
  • Expansion
    14/23 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 14,50 €
    14,50 €
    1 * 1 ST
       14,50 €
  • Expansion
    34/35 NS
    45/40 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,30 €
    52,30 €
    1 * 1 ST
       52,30 €