Orders LinkContactUsLinkComponent
Iris scissors
Iris scissors
Supplier:   KLS Martin Group
Iris scissors
Supplier:  KLS Martin Group
Certificates
2 Options Available Below
Product Details & Documents
Dissecting scissors made of stainless steel. One model features a TC Gold finish.
  • TC Gold finish: Tungsten Carbide inlays provide an exceptionally long service life
  • Straight or curved available
Product Family Options
Product Information
  • Type
    Length
    Availability
    Price
  • Curved, sharp/sharp
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • Straight, sharp/sharp, TC Gold
    115 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST