Orders LinkContactUsLinkComponent
Extractor heads for specific light solvents
Extractor heads for specific light solvents
Supplier:   Lenz Laborglas
Extractor heads for specific light solvents
Supplier:  Lenz Laborglas
Certificates
Extractor heads for specific light solvents
Extractor heads for specific light solvents
3 Options Available Below
Product Details & Documents
DURAN®, Borosilicate glass.
  • Very good chemical resistance
  • Very good temperature resistance
For the extraction of liquids with specific light solvents, with insert filter (porosity: 0).
Documents
Product Family Options
Product Information
  • Extraction cap.
    Availability
    Price
  • 100 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 260,00 €
    260,00 €
    1 * 1 ST
       260,00 €
  • 250 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 302,00 €
    302,00 €
    1 * 1 ST
       302,00 €
  • 500 ml
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 362,00 €
    362,00 €
    1 * 1 ST
       362,00 €