Order Entry
Belgium
ContactUsLinkComponent
Multi-parameter meters, bench, C30xx series
Multi-parameter meters, bench, C30xx series
Catalog # CONSC3050T
Supplier:  CONSORT
undefined
Multi-parameter meters, bench, C30xx series
Catalog # CONSC3050T
Supplier:  CONSORT

Specifications

  • Temperature range:
    −5...+105 °C
  • Description:
    C3050T, kit incl. electrodes (pH/ATC SP21T, ORP SP60X, conductivity SK20T), 2×50 ml buffers (pH 4, 7), 50 ml conductivity standard (0,01 M KCl), 50 ml electrolyte (3 M KCl), flexible electrode holder and cable)
  • Model:
    C3050
  • ISE accuracy:
    -
  • Temperature accuracy:
    0,1 °C
  • Temperature resolution:
    0,1 °C
  • Conductivity accuracy:
    0,5% f.s. of range
  • Conductivity calibration:
    1 - 3 point
  • Conductivity cell constant:
    0,1/1/10 cm⁻¹ ±30%
  • Conductivity range:
    0 - 2000 mS/cm
  • Conductivity reference temperature:
    20/25 °C
  • Conductivity resolution:
    0,001 μS/cm
  • Connections:
    3 BNC, 3x2 banana for Pt1000
  • DO barometric pressure range:
    -
  • DO calibration:
    -
  • DO concentration:
    -
  • DO concentration resolution:
    -
  • DO accuracy:
    -
  • Data logging capacity:
    12000 data sets
  • Display:
    LCD, backlight
  • ISE calibration:
    -
  • ISE resolution:
    -
  • ISE range:
    -
  • Outputs:
    RS232, USB
  • Power supply:
    100 - 240 VAC, 50/60 Hz, low voltage: 9 - 15 VDC
  • Resistivity range:
    0 - 200 MΩ/cm
  • Salinity range:
    0 - 70 ppt
  • Salinity resolution:
    0,1 ppt
  • TDS range:
    0 - 100 g/l
  • TDS resolution:
    0,01 mg/l
  • Temperature calibration:
    1 point
  • Temperature compensation:
    −5...+105 °C
  • Weight:
    1 kg
  • W×D×H:
    260×180×90 mm
  • mV accuracy:
    0,1% ±1 digit
  • mV range:
    ±2000
  • mV resolution:
    0,1
  • pH accuracy:
    0,1% ±1 digit
  • pH calibration:
    1 - 5 point
  • pH range:
    −2...+16
  • pH resolution:
    0,001
  • Pk:
    1

Specifications

About this item

Robust, dust- and splash-proof benchtop meters for several parameters.

  • Large bright LCD screen with white backlight enables to view all channels simultaneously
  • Multi-point (1 - 5) calibration of pH electrodes with any of eleven pre-programmed and five user specified buffer tables
  • Multi-point (1 - 3) calibration of conductivity electrodes for increased linearity with any of three pre-programmed or three user specified standard tables
  • Storage memory for up to 12000 values, date and time
  • Two-way communication with any computer using USB, RS232 interface for connection to a printer

C3010 - pH, mV, conductivity, resistivity, salinity, TDS and temperature
- Two independent channels for all measurements

C3030 - pH, ion, mV, conductivity, resistivity, salinity, TDS, dissolved oxygen and temperature
- Two independent channels for all measurements
- Multi-point (2 - 5) calibration + blank correction of ion-selective electrodes
- Operates with a galvanic oxygen electrode requiring no polarisation time

C3050 - pH, redox, rH2, conductivity, resistivity, salinity, TDS and temperature
- Three independent channels for all measurements (conductivity: two channels)
- Microprocessor controlled bio-electronic rH₂ meter for the study of the biological water quality or illnesses in body fluids according to Vincent's method
- Shows mV referred to the standard hydrogen electrode
- Allows simultaneous measurement of all parameters in the same solution

rH₂ (measurement for the level of electronic exchanges between water and dissolved ions)
Range: 0...42 rH₂
Resolution: 0,01 rH₂
Accuracy: 0,1% ± 1 digit