Orders LinkContactUsLinkComponent
Flasks, round bottom, with three parallel necks
Supplier:   witeg Labortechnik
Flasks, round bottom, with three parallel necks
Supplier:  witeg Labortechnik
Certificates
Flasks, round bottom, with three parallel necks
Flasks, round bottom, with three parallel necks
3 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Typical applications: distillation, extraction.
Product Family Options
Product Information
  • Capacity
    Centre neck
    Side necks
    Availability
    Price
  • 6000 ml
    29/32 NS
    29/32 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 337,00 €
    337,00 €
    1 * 1 ST
       337,00 €
  • 1000 ml
    29/32 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 69,30 €
    69,30 €
    1 * 1 ST
       69,30 €
  • 2000 ml
    29/32 NS
    19/26 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 123,00 €
    123,00 €
    1 * 1 ST
       123,00 €