Orders LinkContactUsLinkComponent
Flasks, round bottom, with two necks
Flasks, round bottom, with two necks
Supplier:   witeg Labortechnik
Flasks, round bottom, with two necks
Supplier:  witeg Labortechnik
Certificates
Flasks, round bottom, with two necks
Flasks, round bottom, with two necks
3 Options Available Below
Product Details & Documents
Borosilicate glass 3.3.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
One vertical centre neck and one angled side neck (20°). With matt white labelling area.
Certifications: DIN 12394
Product Family Options
Product Information
  • Capacity
    Centre neck
    Side necks
    Availability
    Price
  • 100 ml
    14/23 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,60 €
    27,60 €
    1 * 1 ST
       27,60 €
  • 1000 ml
    24/29 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,30 €
    52,30 €
    1 * 1 ST
       52,30 €
  • 100 ml
    24/29 NS
    14/23 NS
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 28,30 €
    28,30 €
    1 * 1 ST
       28,30 €