ContactUsLinkComponent
VWR® Ready Ladder™ DNA Ladder, 50 bp
Catalog # N746-100RXN
Supplier:  VWR Chemicals
CAS Number:  
undefined
VWR® Ready Ladder™ DNA Ladder, 50 bp
Catalog # N746-100RXN
Supplier:  VWR Chemicals
CAS Number:  
Specifications
  • Pk:
    1 KIT
  • Description:
    50 bp Ready Ladder™, pre-mixed with loading dye, sufficient for 100 lanes
Specifications
About this item
Ladder with 10 fragments ranging from 50 to 500 bp for easy band identification. Ready to use formula pre-mixed with loading dye.