Orders LinkContactUsLinkComponent
Adapters, Combifix®
Adapters, Combifix®
Adapters, Combifix®
Supplier:  B. Braun
Certificates
Adapters, Combifix®
Adapters, Combifix®
2 Options Available Below
Product Details & Documents
These adapters are used for connecting fittings.
Suitable for interconnecting male and female fittings with 6% (Luer) and 10% (Rekord) taper fittings.
Documents
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 720-2583
    Luer-Lock (female/female) adapters
    Luer-Lock (female/female) adapters
    Transparent
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 322,00 €
    322,00 €
    1 * 100 ST
       322,00 €
  • BRAU5206642
    Luer-Lock (male/male) adapters
    Luer-Lock (male/male) adapters
    Transparent
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 312,00 €
    312,00 €
    1 * 100 ST
       312,00 €