Orders LinkContactUsLinkComponent
Adapters, reduction, Preciso
Supplier:   witeg Labortechnik
Adapters, reduction, Preciso
Supplier:  witeg Labortechnik
Certificates
Adapters, reduction, Preciso
Adapters, reduction, Preciso
2 Options Available Below
Product Details & Documents
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Reduction adapter from large cone at the base to small socket on the top.
 : According to DIN 12257.
Product Family Options
Product Information
  • Type
    Cone size
    Socket
    Availability
    Price
  • Reduction
    29/32 NS
    19/26
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 9,60 €
    9,60 €
    1 * 1 ST
       9,60 €
  • Reduction
    45/40 NS
    34/35
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 52,30 €
    52,30 €
    1 * 1 ST
       52,30 €