Orders LinkContactUsLinkComponent
Stopcocks, needle-valve, angled
Stopcocks, needle-valve, angled
0 - 6 mm
Catalog # WITG0.243.250
Supplier:   witeg Labortechnik
Stopcocks, needle-valve, angled
Catalog # WITG0.243.250
Supplier:  witeg Labortechnik
Stopcocks, needle-valve, angled
Stopcocks, needle-valve, angled
0 - 6 mm
Price
26,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Needle valve stopcock, Duran, with PTFE-needle valve, sealing system and overtwist safety device.
  • Very good chemical resistance
  • Very good temperature resistance
Specifications
  • Hole Ø:
    0 - 6 mm