ContactUsLinkComponent
Spectrophotometric colorimeters, model F, Lovibond®
Spectrophotometric colorimeters, model F, Lovibond®
Certificates
undefined
Spectrophotometric colorimeters, model F, Lovibond®
Specifications
  • Catalog No:
  • TINT180270
  • 633-0138
  • Model:
  • Model F
  • Model F
  • Light source:
  • 2×12 Volt, 10 W tungsten halogen lamp
  • 2×12 Volt, 10 W tungsten halogen lamp
  • Illuminants:
  • Approximates to daylight
  • Approximates to daylight
  • Optical design:
  • Visual comparison of sample with colour standards in transmittance and reflectance modes
  • Visual comparison of sample with colour standards in transmittance and reflectance modes
  • Path length:
  • 0,1 - 153 mm
  • 0,1 - 153 mm
  • W×D×H:
  • 330×410×230 mm
  • 330×410×230 mm
  • Weight:
  • 8,3 kg
  • 8,3 kg
Specifications
Spare Colour Filter Racks
Product Information
Frequently Bought Together