ContactUsLinkComponent
Magnetic stirring bars, double cross-head, PTFE
Magnetic stirring bars, double cross-head, PTFE
Catalog # COWI001.1135
Supplier:  COWIE
Magnetic stirring bars, double cross-head, PTFE
Catalog # COWI001.1135
Supplier:  COWIE
Specifications
  • Ø:
    35 mm
  • Height:
    12 mm
  • Pk:
    1
Specifications
About this item
Double cross-head has a greater turbulance when compared to single cross-head.