ContactUsLinkComponent
Rotating shaker, Rotator Genie™
Rotating shaker, Rotator Genie™
Certificates
undefined
Rotating shaker, Rotator Genie™
Specifications
  • Catalog No:
  • SCIISI-2102
  • SCIISI-2202
  • Model:
  • Rotator Genie, standard platform
  • Rotator Genie with magnetic platform
  • Shaking motion:
  • Rotating
  • Rotating
  • Speed:
  • 5 - 35 min⁻¹
  • 5 - 35 min⁻¹
  • Angle:
  • Rotates 360 degrees, standard platform can be turned 90 degrees
  • Rotates 360 degrees, standard platform can be turned 90 degrees
  • W×D×H:
  • 175×400×400 mm
  • 175×400×400 mm
  • Platform W×D:
  • 260×260 mm
  • 290×315 mm
  • Weight:
  • 5,2 kg
  • 5,2 kg
  • Max. load:
  • 1,8 kg
  • 1,8 kg