ContactUsLinkComponent
Neutralisation agent, Helimatic® Neutralizer C
Neutralisation agent, Helimatic® Neutralizer C
Catalog # BRAU18732
Supplier:  B. Braun
undefined
Neutralisation agent, Helimatic® Neutralizer C
Catalog # BRAU18732
Supplier:  B. Braun
Specifications
  • Capacity:
    5 l
  • Description:
    Helimatic® Neutralizer C (based on citric acid)
  • Pk:
    5 kg
Specifications
About this item
Helimatic® Neutralizer C is a liquid neutraliser containing citric acid which is used for the neutralisation of alkaline resides during machine processing of instruments. It has a low working concentration from 0,05% - 0,3%.
  • Automatic reprocessing of resuable invasive and non invasive medical devices
  • Free of phosphates and surfactants
  • Based on citric acid (pharmaceutical grade)