Order Entry
Belgium
Orders LinkContactUsLinkComponent
Uridine

Uridine

Glass bottle for solids, 25 g
Catalog # MOLEM20959741
Marketsource
Supplier:   Molekula
Uridine
Catalog # MOLEM20959741
Supplier:  Molekula
Marketsource
CAS Number:  58-96-8
Uridine

Glass bottle for solids, 25 g

CAS Number:  58-96-8
Price
Inquire For Price
Log in for availability
1 * 25 g
Product Details & Documents
Synonyms:  1-((2R,3R,4S,5R)-3,4-Dihydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)pyrimidine-2,4(1H,3H)-dione, 1-β-D-Ribofuranosyluracil, Uracil-1-β-D-ribofuranoside
  • CAS Number:58-96-8
  • Formula:C₉H₁₂N₂O₆
  • MDL Number:MFCD00006526
  • Molecular Weight:244.20 g/mol
  • Storage Temperature:Ambient
  • EINECS:200-407-5
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.