Order Entry
Belgium
ContactUsLinkComponent
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Irritant
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # MOLE11939321-500MG
Supplier:  Molekula
CAS Number:  
Irritant
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # MOLE11939321-500MG
Supplier:  Molekula
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 mg
  • Seq:
    2725

Specifications

About this item

 :
 :239.69 g/mol
Storage Temperature:Refrigerator
UN:3261
ADR:8,II
 :GHS07
CAS Number:30827-99-7
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962