Order Entry
Belgium
ContactUsLinkComponent
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # 786-053
Supplier:  G-Biosciences
CAS Number:  
AEBSF hydrochloride (4-(2-aminoethyl)benzenesulphonylfluoride hydrochloride)
Catalog # 786-053
Supplier:  G-Biosciences
CAS Number:  

Specifications

  • Pk:
    1 g
  • Seq:
    3047

Specifications

About this item

 :
 :239.69 g/mol
ADR:8,II
CAS Number:30827-99-7
Formula:C₈H₁₁ClFNO₂S
MDL Number:MFCD00132962