
Lithium chloride 8 mol/l in aqueous solution
Catalog # SERA39561.02
Supplier: Serva
CAS Number:
Specifications
- Pack type:Glass bottle
- Pk:1 L
- Seq:5037
Specifications
About this item
:
:42.39 g/mol
EINECS:231-212-3
:GHS07
Storage Temperature:Ambient
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078