Order Entry
Belgium
ContactUsLinkComponent
Lithium chloride 8 mol/l in aqueous solution for biotechnology
Irritant
Lithium chloride 8 mol/l in aqueous solution for biotechnology
Catalog # K445-100ML
Supplier:  VWR Chemicals
CAS Number:  
undefined
Irritant
Lithium chloride 8 mol/l in aqueous solution for biotechnology
Catalog # K445-100ML
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    100 mL
  • Seq:
    2440

Specifications

Specification Test Results

  • DNase:
    None Detected
  • Protease:
    None Detected
  • RNase:
    None Detected
  • Titration:
    7.7 – 8.2 M
  • Appearance:
    Clear, colorless liquid

Specification Test Results

About this item

 :
 :42.39 g/mol
EINECS:231-212-3
 :GHS07
Storage Temperature:Ambient
CAS Number:7447-41-8
Formula:LiCl
MDL Number:MFCD00011078