Order Entry
Belgium
ContactUsLinkComponent
Ribonuclease A
Ribonuclease A
Supplier:  PanReac AppliChem
Certificates
Ribonuclease A
Supplier:  PanReac AppliChem
Certificates

Specifications

  • CAS No.:
    9001-99-4
  • Storage conditions:
    Freezer
  • MDL No.:
    MFCD00132184
  • Enzyme name:
    Ribonuclease A
  • Enzyme synonyms:
    Rnase A

Specifications

Related Information

Frequently Bought Together