Order Entry
Belgium
ContactUsLinkComponent
Ethanol denatured absolute ≥98% (by GC) (denaturated with 1% MEK and 0.001% Bitrex®)
Irritant
Flammable
Ethanol denatured absolute ≥98% (by GC) (denaturated with 1% MEK and 0.001% Bitrex®)
Certificates
Irritant
Flammable
Ethanol denatured absolute ≥98% (by GC) (denaturated with 1% MEK and 0.001% Bitrex®)
Certificates

Specification Test Results

  • Identity:
    complying
  • Assay (GC, without denaturant):
    Min. 99.8 %
  • Water (Karl Fischer):
    Max. 0.2 %
  • Free acid (as CH₃COOH):
    Max. 0.002 %
  • methyl ethyl ketone (GC):
    1.0 - 1.5 Vol.%
  • 2-propanol (GC):
    1.0 - 1.5 Vol.%
  • Bitrex min. 1,0 g/100 l:
    complying
  • Autoignition Temperature:
    683 °F
  • Explosion Limit:
    19 %, 60 °F
  • Vapor Pressure:
    44.6 mmHg ( 20 °C)
  • Vapor Density:
    1.59 (vs air)

Specification Test Results

Related Information

Frequently Bought Together