Order Entry
Belgium
ContactUsLinkComponent
Sodium formate ≥99% ACS
Sodium formate ≥99% ACS
Catalog # J961-100G
Supplier:  VWR Chemicals
CAS Number:  
undefined
Sodium formate ≥99% ACS
Catalog # J961-100G
Supplier:  VWR Chemicals
CAS Number:  

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    100 g
  • Seq:
    3401

Specifications

Specification Test Results

  • Calcium:
    <0.005%
  • Chloride:
    ≤0.001%
  • Heavy Metals:
    ≤0.0005%
  • Insolubles:
    ≤0.005%
  • Iron:
    ≤0.0005%
  • Purity:
    ≥99%
  • Sulphate:
    ≤0.001%

Specification Test Results

About this item

 :
 :68.01 g/mol
Merck Index:13,08694
Melting Pt:257.31 °C
Density:1.919 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-488-0
REACH:01-2119486468-21
CAS Number:141-53-7
Formula:HCO₂Na
MDL Number:MFCD00013101